3,5-Bis(trifluoromethyl)phenol 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
α,α,α,α',α',α'-Hexafluoro-3,5-xylenol
PC3112-100G
PC3112-25G
APOSPC3112-100GEA
132
EUR
APOSPC3112-100G
APOSPC3112-25G
3,5-Bis(trifluoromethyl)phenol 98%
3,5-Bis(trifluoromethyl)phenol
Formula:
C₈H₄F₆O Boiling Pt: 97…98 °C/50mmHg Melting Pt: 20…21 °C Density: 1.511 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000386 CAS Number: 349-58-6 EINECS: 206-488-3 |