3,5-Bis(trifluoromethyl)phenylacetonitrile 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(3,5-Bis(trifluoromethyl)phenyl)acetonitrile
PC3114G-5G PC3114G-100G PC3114G-25G
APOSPC3114G-5GEA 21.9 EUR
APOSPC3114G-5G APOSPC3114G-100G APOSPC3114G-25G
3,5-Bis(trifluoromethyl)phenylacetonitrile 95%
3,5-Bis(trifluoromethyl)phenylacetonitrile
Formula: C₁₀H₅F₆N
Density: 1.42 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009904
CAS Number: 85068-32-2
EINECS: 285-294-0
UN: 3276
ADR: 6.1,III