1-(3,5-Bis(trifluoromethyl)phenyl)thiourea 98%
Supplier: Apollo Scientific
Danger
Synonyms: N-[3,5-Bis(trifluoromethyl)phenyl]thiourea
PC3115M-1G PC3115M-5G
APOSPC3115M-1GEA 61.1 EUR
APOSPC3115M-1G APOSPC3115M-5G
1-(3,5-Bis(trifluoromethyl)phenyl)thiourea 98%
1-(3,5-Bis(trifluoromethyl)phenyl)thiourea
Formula: C₉H₆F₆N₂S
MW: 288.22 g/mol
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00041172
CAS Number: 175277-17-5
UN: 2811
ADR: 6.1,III