3',5'-Bis(trifluoromethyl)propiophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3,5-Bis(trifluoromethyl)phenyl)propan-1-one
PC3116-5G PC3116-25G PC3116-100G
APOSPC3116-5GEA 21.9 EUR
APOSPC3116-5G APOSPC3116-25G APOSPC3116-100G
3',5'-Bis(trifluoromethyl)propiophenone 98%
3',5'-Bis(trifluoromethyl)propiophenone
Formula: C₁₁H₈F₆O
MW: 270.17 g/mol
Boiling Pt: 72 °C/7mmHg
Density: 1.379 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00009911
CAS Number: 85068-34-4
EINECS: 285-296-1