3-Fluorobenzenesulphonamide 97%
Supplier: Apollo Scientific
Danger
PC3117-25G PC3117-5G
APOSPC3117-25GEA 230 EUR
APOSPC3117-25G APOSPC3117-5G
3-Fluorobenzenesulphonamide 97%
3-Fluorobenzenesulphonamide
Formula: C₆H₆FNO₂S
MW: 175.18 g/mol
Melting Pt: 124…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00042284
CAS Number: 1524-40-9
UN: 2811
ADR: 6.1,III