4-Amino-3-(trifluoromethyl)benzonitrile 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-5-cyanobenzotrifluoride
PC31183-100G PC31183-25G
APOSPC31183-100GEA 132 EUR
APOSPC31183-100G APOSPC31183-25G
4-Amino-3-(trifluoromethyl)benzonitrile 99%
4-Amino-3-(trifluoromethyl)benzonitrile
Formula: C₈H₅F₃N₂
MW: 186.14 g/mol
Boiling Pt: 100 °C/0.1mmHg
Melting Pt: 60…63 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00275473
CAS Number: 327-74-2
UN: 3439
ADR: 6.1 III