2-[N,N-Bis(trifluoromethanesulphonyl)amino]-5-chloropyridine
Supplier: Apollo Scientific
Warning
PC3128-25G PC3128-100G PC3128-5G
APOSPC3128-25GEA 78.3 EUR
APOSPC3128-25G APOSPC3128-100G APOSPC3128-5G
2-[N,N-Bis(trifluoromethanesulphonyl)amino]-5-chloropyridine
Formula: C₇H₃ClF₆N₂O₄S₂
Melting Pt: 45…47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191833
CAS Number: 145100-51-2