5-Chloro-2-fluorobenzoic acid 97%
Supplier: Apollo Scientific
Warning
PC3142-100G PC3142-25G PC3142-5G PC3142-500G
APOSPC3142-100GEA 62.5 EUR
APOSPC3142-100G APOSPC3142-25G APOSPC3142-5G APOSPC3142-500G
5-Chloro-2-fluorobenzoic acid 97%
5-Chloro-2-fluorobenzoic acid
Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
Melting Pt: 148…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00665762
CAS Number: 394-30-9