2,4-Dimethoxy-5-(trifluoromethyl)aniline
Supplier: Apollo Scientific
Warning
PC31485-1G PC31485-5G
APOSPC31485-1GEA 145 EUR
APOSPC31485-1G APOSPC31485-5G
2,4-Dimethoxy-5-(trifluoromethyl)aniline
2,4-Dimethoxy-5-(trifluoromethyl)aniline
Formula: C₉H₁₀F₃NO₂
Melting Pt: 78…80 °C
Storage Temperature: Ambient
CAS Number: 228401-47-6