1H,1H,11H-Eicosafluoroundecyl acrylate 95%
Supplier: Apollo Scientific
Warning
PC3154-100G PC3154-25G PC3154-5G
APOSPC3154-100GEA 800 EUR
APOSPC3154-100G APOSPC3154-25G APOSPC3154-5G
1H,1H,11H-Eicosafluoroundecyl acrylate 95%
1H,1H,11H-Eicosafluoroundecyl acrylate
Formula: C₁₄H₆F₂₀O₂
Boiling Pt: 115 °C/1mmHg
Melting Pt: 44…45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042112
CAS Number: 4998-38-3
EINECS: 225-659-3