8-(Chloromethyl)-6-fluorobenzo-1,3-dioxane 97%
Supplier: Apollo Scientific
PC31548-1G PC31548-250MG
APOSPC31548-1GEA 98.9 EUR
APOSPC31548-1G APOSPC31548-250MG
8-(Chloromethyl)-6-fluorobenzo-1,3-dioxane 97%
8-(Chloromethyl)-6-fluorobenzo-1,3-dioxane
Formula: C₉H₈ClFO₂
Melting Pt: 81…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00052603
CAS Number: 131728-94-4