1,2-Epoxy-3-fluoropropane 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Fluoro-1,2-propenoxide
PC3158-1G PC3158-25G PC3158-5G
APOSPC3158-1GEA 102 EUR
APOSPC3158-1G APOSPC3158-25G APOSPC3158-5G
1,2-Epoxy-3-fluoropropane 98%
1,2-Epoxy-3-fluoropropane
Formula: C₃H₅FO
MW: 76.07 g/mol
Boiling Pt: 85…86 °C
Density: 1.067 g/cm³ (25 °C)
Flash Pt: 4 °C
Storage Temperature: Refrigerator
CAS Number: 503-09-3
EINECS: 207-960-1
UN: 2924
ADR: 3+8,II