Ethyl-3-amino-4,4,4-trifluorocrotonate 97%
Supplier: Apollo Scientific
Danger
Synonyms: Ethyl 3-amino-4,4,4-trifluorobut-2-enoate
PC3161-100G PC3161-250G PC3161-25G PC3161-500G
APOSPC3161-100GEA 55.1 EUR
APOSPC3161-100G APOSPC3161-250G APOSPC3161-25G APOSPC3161-500G
Ethyl-3-amino-4,4,4-trifluorocrotonate 97%
Ethyl 3-amino-4,4,4-trifluorocrotonate
Formula: C₆H₈F₃NO₂
MW: 183.13 g/mol
Boiling Pt: 83…85 °C/15mmHg
Melting Pt: 24…26 °C
Density: 1.245 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00068195
CAS Number: 372-29-2
UN: 3259
ADR: 8,III