Ethyl-2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate
PC3161E-1G PC3161E-5G
APOSPC3161E-1GEA 26.1 EUR
APOSPC3161E-1G APOSPC3161E-5G
Ethyl-2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate
Ethyl-2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate
Formula: C₈H₈F₃N₃O₂
MW: 235.17 g/mol
Melting Pt: 179…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00052070
CAS Number: 149771-09-5