2-Ethoxy-6-(2,2,2-trifluoroethoxy)benzonitrile, Technical Grade
Supplier: Apollo Scientific
PC3166-1G PC3166-5G PC3166-10G
APOSPC3166-1GEA 72.5 EUR
APOSPC3166-1G APOSPC3166-5G APOSPC3166-10G
2-Ethoxy-6-(2,2,2-trifluoroethoxy)benzonitrile, Technical Grade
2-Ethoxy-6-(2,2,2-trifluoroethoxy)benzonitrile
Formula: C₁₁H₁₀F₃NO₂
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00068218
CAS Number: 175204-04-3