3-Bromo-2-fluorobenzylalkohol 98%

Supplier: Apollo Scientific
Warning

APOSPC3167-1GEA 14.7 EUR
APOSPC3167-1G
3-Bromo-2-fluorobenzylalkohol 98%
3-Bromo-2-fluorobenzylalkohol
Formula: C₇H₆BrFO
MW: 205,0200 g/mol
Boiling Pt: 104…108 °C/4mmHg
Storage Temperature: Ambient
MDL Number: MFCD09842441
CAS Number: 261723-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR