Ethyl-2-(4-chlorophenyl)-3-(trifluoromethyl)pyrazole-4-carboxylate 97%
Supplier: Apollo Scientific
Warning
PC3184-1G PC3184-250MG
APOSPC3184-1GEA 87 EUR
APOSPC3184-1G APOSPC3184-250MG
Ethyl-2-(4-chlorophenyl)-3-(trifluoromethyl)pyrazole-4-carboxylate 97%
Ethyl-2-(4-chlorophenyl)-3-(trifluoromethyl)pyrazole-4-carboxylate
Formula: C₁₃H₁₀ClF₃N₂O₂
MW: 318.69 g/mol
Boiling Pt: 126…128 °C/0.02mmHg
Melting Pt: 52…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00052074
CAS Number: 112055-36-4