1-Chloro-4,5-difluoro-2-nitrobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-4,5-difluoronitrobenzene
PC3191-1G PC3191-25G PC3191-5G
APOSPC3191-1GEA 21.9 EUR
APOSPC3191-1G APOSPC3191-25G APOSPC3191-5G
1-Chloro-4,5-difluoro-2-nitrobenzene 98%
1-Chloro-4,5-difluoro-2-nitrobenzene
Formula: C₆H₂ClF₂NO₂
MW: 193.5354 g/mol
Boiling Pt: 118 °C/31mmHg
Storage Temperature: Ambient
MDL Number: MFCD04973764
CAS Number: 771-76-6