Methyl-4-chloro-3-fluorobenzoate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 4-chloro-3-fluorobenzoate, 4-Chloro-3-fluorobenzoic acid methyl ester
PC31950-1G PC31950-25G PC31950-5G
APOSPC31950-1GEA 21.9 EUR
APOSPC31950-1G APOSPC31950-25G APOSPC31950-5G
Methyl-4-chloro-3-fluorobenzoate
Methyl 4-chloro-3-fluorobenzoate
Formula: C₈H₆ClFO₂
MW: 188.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00209299
CAS Number: 206362-87-0