Ethyl 2,4-difluorobenzoate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 2,4-difluorobenzoate
PC3201E-25G PC3201E-100G
APOSPC3201E-25GEA 23.2 EUR
APOSPC3201E-25G APOSPC3201E-100G
Ethyl 2,4-difluorobenzoate 97%
Ethyl 2,4-difluorobenzoate
Formula: C₉H₈F₂O₂
MW: 186.16 g/mol
Boiling Pt: 65…67 °C/15mmHg
Density: 1.24 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00153149
CAS Number: 108928-00-3