8-Fluoroquinoline
Supplier: Apollo Scientific
Warning
PC3205-1G PC3205-25G PC3205-5G
APOSPC3205-1GEA 21.9 EUR
APOSPC3205-1G APOSPC3205-25G APOSPC3205-5G
8-Fluoroquinoline
8-Fluoroquinoline
Formula: C₉H₆FN
MW: 147.1500 g/mol
Boiling Pt: 148…150 °C/30mmHg
Density: 1.215 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01685514
CAS Number: 394-68-3