3-Bromo-4-(trifluoromethoxy)aniline 98%
Supplier: Apollo Scientific
|
Warning
|
PC3215-100G
PC3215-25G
PC3215-5G
APOSPC3215-100GEA
115
EUR
APOSPC3215-100G
APOSPC3215-25G
APOSPC3215-5G
3-Bromo-4-(trifluoromethoxy)aniline 98%
3-Bromo-4-(trifluoromethoxy)aniline
Formula:
C₇H₅BrF₃NO MW: 256.02 g/mol Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190127 CAS Number: 191602-54-7 UN: 2810 ADR: 6.1 III |