4-Fluoro-3-nitrobenzaldehyde
Supplier: Apollo Scientific
|
Warning
|
PC32290-100G
PC32290-25G
PC32290-5G
APOSPC32290-100GEA
132
EUR
APOSPC32290-100G
APOSPC32290-25G
APOSPC32290-5G
4-Fluoro-3-nitrobenzaldehyde
4-Fluoro-3-nitrobenzaldehyde
Formula:
C₇H₄FNO₃ MW: 169.11 g/mol Melting Pt: 45…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD01861388 CAS Number: 42564-51-2 |