4-Chloro-2-(trifluoromethoxy)aniline 95%
Supplier: Apollo Scientific
Warning
PC32387-1G PC32387-5G PC32387-10G
APOSPC32387-1GEA 24.8 EUR
APOSPC32387-1G APOSPC32387-5G APOSPC32387-10G
4-Chloro-2-(trifluoromethoxy)aniline 95%
4-Chloro-2-(trifluoromethoxy)aniline
Formula: C₇H₅ClF₃NO
Boiling Pt: 82 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00153102
CAS Number: 175205-77-3
UN: 2810
ADR: 6.1,III