2-Bromo-4-(trifluoromethyl)pyridine 95%
Supplier: Apollo Scientific
Warning
PC32398-25G PC32398-5G
APOSPC32398-25GEA 157 EUR
APOSPC32398-25G APOSPC32398-5G
2-Bromo-4-(trifluoromethyl)pyridine 95%
2-Bromo-4-(trifluoromethyl)pyridine
Formula: C₆H₃BrF₃N
MW: 226 g/mol
Boiling Pt: 84…85 °C/14mmHg
Melting Pt: –21…–20 °C
Density: 1.827 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00153085
CAS Number: 175205-81-9