2-(4-Trifluoromethylpyridin-2-yloxy)ethanol
Supplier: Apollo Scientific
Warning
Synonyms: 2-(2-Hydroxyethoxy)-4-(trifluoromethyl)pyridine
PC32400-1G PC32400-250MG
APOSPC32400-1GEA 328 EUR
APOSPC32400-1G APOSPC32400-250MG
2-(4-Trifluoromethylpyridin-2-yloxy)ethanol
2-(4-Trifluoromethylpyridin-2-yloxy)ethanol
Formula: C₈H₈F₃NO₂
MW: 207.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00126446
CAS Number: 257295-59-3
UN: 2811
ADR: 6.1,III