3-Amino-2-methylbenzotrifluoride 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Methyl-3-(trifluoromethyl)aniline
PC3243-100G PC3243-25G PC3243-5G PC3243-500G
APOSPC3243-100GEA 81.2 EUR
APOSPC3243-100G APOSPC3243-25G APOSPC3243-5G APOSPC3243-500G
3-Amino-2-methylbenzotrifluoride 98%
3-Amino-2-methylbenzotrifluoride
Formula: C₈H₈F₃N
MW: 175.16 g/mol
Boiling Pt: 62…64 °C/4mmHg
Melting Pt: 37…42 °C
Density: 1.3 g/cm³ (25 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00153216
CAS Number: 54396-44-0
EINECS: 259-145-5