Ethyl-4,4,4-trifluoro-3-hydroxybutyrate 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4,4,4-Trifluoro-3-hydroxybutyric Acid Ethyl Ester, Ethyl 4,4,4-trifluoro-3-hydroxybutyrate
PC3244-25G PC3244-100G
APOSPC3244-25GEA 91.5 EUR
APOSPC3244-25G APOSPC3244-100G
Ethyl-4,4,4-trifluoro-3-hydroxybutyrate 97%
Ethyl-4,4,4-trifluoro-3-hydroxybutyrate
Formula: C₆H₉F₃O₃
MW: 186.13 g/mol
Boiling Pt: 90…91 °C/50mmHg
Melting Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00040842
CAS Number: 372-30-5