2,3-Difluoro-4-(trifluoromethyl)benzoic acid 98%
Supplier: Apollo Scientific
Warning
PC3249-1G PC3249-5G
APOSPC3249-1GEA 197 EUR
APOSPC3249-1G APOSPC3249-5G
2,3-Difluoro-4-(trifluoromethyl)benzoic acid 98%
2,3-Difluoro-4-(trifluoromethyl)benzoic acid
Formula: C₈H₃F₅O₂
Melting Pt: 119…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00236256
CAS Number: 237424-17-8