Ethyl pentafluoropropionylacetate
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 3-oxo-4,4,5,5,5-pentafluoropentanoate
PC3250F-25G PC3250F-5G PC3250F-1G
APOSPC3250F-25GEA 212 EUR
APOSPC3250F-25G APOSPC3250F-5G APOSPC3250F-1G
Ethyl pentafluoropropionylacetate
Ethyl pentafluoropropionylacetate
Formula: C₇H₇F₅O₃
MW: 234.12 g/mol
Boiling Pt: 141…145 °C
Density: 1.338 g/cm³ (25 °C)
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00013569
CAS Number: 663-35-4
UN: 3272
ADR: 3 III