5-Amino-2,2-difluoro-1,3-benzodioxole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2,2-Difluoro-1,3-benzodioxol-5-amine, 3,4-(Difluoromethylenedioxy)aniline
PC3252-1G PC3252-5G PC3252-10G
APOSPC3252-1GEA 21.9 EUR
APOSPC3252-1G APOSPC3252-5G APOSPC3252-10G
5-Amino-2,2-difluoro-1,3-benzodioxole 98%
5-Amino-2,2-difluoro-1,3-benzodioxole
Formula: C₇H₅F₂NO₂
MW: 173.12 g/mol
Boiling Pt: 95…97 °C/12mmHg
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00190144
CAS Number: 1544-85-0
UN: 2810
ADR: 6.1 III