2-Fluoro-3-iodobenzoic acid
Supplier: Apollo Scientific
Warning
PC32730-1G PC32730-25G PC32730-5G
APOSPC32730-1GEA 37.7 EUR
APOSPC32730-1G APOSPC32730-25G APOSPC32730-5G
2-Fluoro-3-iodobenzoic acid
2-Fluoro-3-iodobenzoic acid
Formula: C₇H₄FIO₂
MW: 266.0083 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07774248
CAS Number: 447464-03-1