tert-Butyl-4-bromo-2-fluorobenzoate 98%
Supplier: Apollo Scientific
Synonyms: tert-Butyl 4-bromo-2-fluorobenzoate
PC32732-25G PC32732-5G
APOSPC32732-25GEA 157 EUR
APOSPC32732-25G APOSPC32732-5G
tert-Butyl-4-bromo-2-fluorobenzoate 98%
tert-Butyl 4-bromo-2-fluorobenzoate
Formula: C₁₁H₁₂BrFO₂
MW: 275.1100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08703426
CAS Number: 889858-12-2