3-Fluoro-4-iodobenzoic acid 98%
Supplier: Apollo Scientific
Warning
PC32733-25G PC32733-5G PC32733-100G
APOSPC32733-25GEA 84.1 EUR
APOSPC32733-25G APOSPC32733-5G APOSPC32733-100G
3-Fluoro-4-iodobenzoic acid 98%
3-Fluoro-4-iodobenzoic acid
Formula: C₇H₄FIO₂
MW: 266.0100 g/mol
Melting Pt: 229…230 °C
Storage Temperature: Ambient
MDL Number: MFCD07774250
CAS Number: 825-98-9