1-Methyl-3-trifluoromethyl-2-pyrazolin-5-one 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,4-Dihydro-2-methyl-5-(trifluoromethyl)-3H-pyrazol-3-one
PC3274-10G PC3274-25G
APOSPC3274-10GEA 60.9 EUR
APOSPC3274-10G APOSPC3274-25G
1-Methyl-3-trifluoromethyl-2-pyrazolin-5-one 97%
1-Methyl-3-trifluoromethyl-2-pyrazolin-5-one
Formula: C₅H₅F₃N₂O
MW: 166.1 g/mol
Melting Pt: 178…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00051654
CAS Number: 1481-02-3