Ethyl-4,4,4-trifluorobutyrate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 4,4,4-trifluorobutanoate, Ethyl 4,4,4-trifluorobutyrate
PC3281-25G
APOSPC3281-25GEA 119 EUR
APOSPC3281-25G
Ethyl-4,4,4-trifluorobutyrate 98%
Ethyl 4,4,4-trifluorobutyrat
Formula: C₆H₉F₃O₂
MW: 170.13 g/mol
Boiling Pt: 127 °C
Density: 1.16 g/cm³ (25 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00041398
CAS Number: 371-26-6
UN: 3272
ADR: 3,III