Ethyl-4,4,4-trifluorocrotonate 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ethyl (E)-4,4,4-trifluorobut-2-enoate, Ethyl 4,4,4-trifluorocrotonate
PC3282-100G
PC3282-25G
APOSPC3282-100GEA
314
EUR
APOSPC3282-100G
APOSPC3282-25G
Ethyl-4,4,4-trifluorocrotonate 98%
Ethyl 4,4,4-trifluorocrotonate
Formula:
C₆H₇F₃O₂ MW: 168.12 g/mol Boiling Pt: 113…115 °C Density: 1.125 g/cm³ (25 °C) Flash Pt: 25 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009903 CAS Number: 25597-16-4 UN: 3272 ADR: 3,III |