Ethyl-3-(trifluoromethyl)crotonate 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Trifluoromethyl)crotonic acid ethyl ester
PC3288C-25G PC3288C-5G
APOSPC3288C-25GEA 400 EUR
APOSPC3288C-25G APOSPC3288C-5G
Ethyl-3-(trifluoromethyl)crotonate 97%
Ethyl-3-(trifluoromethyl)crotonate
Formula: C₇H₉F₃O₂
MW: 182.14 g/mol
Boiling Pt: 129…130 °C
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00040846
CAS Number: 24490-03-7
UN: 3272
ADR: 3 III