Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)crotonate 95%
Supplier: Apollo Scientific
Warning
PC3289G-25G PC3289G-5G
APOSPC3289G-25GEA 694 EUR
APOSPC3289G-25G APOSPC3289G-5G
Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)crotonate 95%
Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)crotonate
Formula: C₇H₆F₆O₂
Boiling Pt: 127…128 °C
Density: 1.3525 g/cm³ (25 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00041517
CAS Number: 1513-60-6
UN: 1993
ADR: 3,III