Iron(II) fluoride, anhydrous 99%
Supplier: Apollo Scientific
Danger
PC3300-25G PC3300-5G
APOSPC3300-25GEA 335 EUR
APOSPC3300-25G APOSPC3300-5G
Iron(II) fluoride, anhydrous 99%
Iron (II) fluoride
Formula: F₂Fe
Melting Pt: >1000 °C
Density: 4.09 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016093
CAS Number: 7789-28-8
UN: 3260
ADR: 8,II