3-Fluoro-2-methylbenzotrifluoride 98%
Supplier: Apollo Scientific
Danger
PC3319-1G PC3319-25G PC3319-5G
APOSPC3319-1GEA 21.9 EUR
APOSPC3319-1G APOSPC3319-25G APOSPC3319-5G
3-Fluoro-2-methylbenzotrifluoride 98%
3-Fluoro-2-methylbenzotrifluoride
Formula: C₈H₆F₄
Density: 1.2 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD08062608
CAS Number: 910911-43-2
UN: 1993
ADR: 3,II