Methyl-4-(trifluoromethyl)pyridine-3-carboxylate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 4-(trifluoromethyl)nicotinate
PC3321-1G PC3321-250MG
APOSPC3321-1GEA 102 EUR
APOSPC3321-1G APOSPC3321-250MG
Methyl-4-(trifluoromethyl)pyridine-3-carboxylate 97%
Methyl-4-(trifluoromethyl)pyridine-3-carboxylate
Formula: C₈H₆F₃NO₂
Boiling Pt: 100 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00101568
CAS Number: 175204-82-7