Perfluoro-1-hexene
Supplier: Apollo Scientific
Warning
PC3341-1G PC3341-5G
APOSPC3341-1GEA 109 EUR
APOSPC3341-1G APOSPC3341-5G
Perfluoro-1-hexene
Perfluoro-1-hexene
Formula: C₆F₁₂
Boiling Pt: 57 °C
Density: 1.3525 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD04038339
CAS Number: 755-25-9