Perfluoro(2-methyl-3-pentanone)
Supplier: Apollo Scientific
Warning
Synonyms: 1,1,1,2,2,4,5,5,5-Nonafluoro-4-(trifluoromethyl)-3-pentanone, Perfluoroethyl Perfluoroisopropyl ketone
PC3347-100G PC3347-25G PC3347-5G
APOSPC3347-100GEA 267 EUR
APOSPC3347-100G APOSPC3347-25G APOSPC3347-5G
Perfluoro(2-methyl-3-pentanone)
Perfluoro(2-methyl-3-pentanone)
Formula: C₆F₁₂O
MW: 316.05 g/mol
Boiling Pt: 49 °C
Melting Pt: –108 °C
Density: 1.6 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD04038343
CAS Number: 756-13-8
EINECS: 436-710-6