2-(Trifluoroacetyl)pyrrole 99%
Supplier: Apollo Scientific
Warning
PC3389-25G PC3389-5G PC3389-1G
APOSPC3389-25GEA 548 EUR
APOSPC3389-25G APOSPC3389-5G APOSPC3389-1G
2-(Trifluoroacetyl)pyrrole 99%
2-(Trifluoroacetyl)pyrrole
Formula: C₆H₄F₃NO
MW: 163.100 g/mol
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00141809
CAS Number: 2557-70-2