4,4,4-Trifluorocrotonic acid 97%
Supplier: Apollo Scientific
Danger
Synonyms: 4,4,4-Trifluorobut-2-enoic acid
PC3392-25G PC3392-5G
APOSPC3392-25GEA 323 EUR
APOSPC3392-25G APOSPC3392-5G
4,4,4-Trifluorocrotonic acid 97%
4,4,4-Trifluorocrotonic acid
Formula: C₄H₃F₃O₂
MW: 140.06 g/mol
Boiling Pt: 68…69 °C/20mmHg
Melting Pt: 55…56 °C
Density: 1.4 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD02181180
CAS Number: 71027-02-6
UN: 3261
ADR: 8,III