4,4,4-Trifluoro-2-iodobut-2-en-1-ol 97%
Supplier: Apollo Scientific
PC3395-25G PC3395-5G PC3395-1G
APOSPC3395-25GEA 746 EUR
APOSPC3395-25G APOSPC3395-5G APOSPC3395-1G
4,4,4-Trifluoro-2-iodobut-2-en-1-ol 97%
4,4,4-Trifluoro-2-iodobut-2-en-1-ol
Formula: C₄H₄F₃IO
MW: 251.98 g/mol
Boiling Pt: 75 °C/10mmHg
Density: 2.076 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00156050
CAS Number: 674-35-1