3-Fluoroaniline 98%
Supplier: Apollo Scientific
Danger
PC3400-100G PC3400-1KG PC3400-500G PC3400-25G
APOSPC3400-100GEA 143 EUR
APOSPC3400-100G APOSPC3400-1KG APOSPC3400-500G APOSPC3400-25G
3-Fluoroaniline 98%
3-Fluoroaniline
Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 186…187 °C
Melting Pt: –2 °C
Density: 1.156 g/cm³ (25 °C)
Flash Pt: 77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007758
CAS Number: 372-19-0
EINECS: 206-747-0
UN: 2941
ADR: 6.1,III