2,4-Dichloro-5-fluorobenzoyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,4-Dichloro-5-fluorobenzene-1-carbonyl chloride
PC3408-25G PC3408-5G
APOSPC3408-25GEA 234 EUR
APOSPC3408-25G APOSPC3408-5G
2,4-Dichloro-5-fluorobenzoyl chloride 98%
2,4-Dichloro-5-fluorobenzoyl chloride
Formula: C₇H₂Cl₃FO
Boiling Pt: 143…144 °C/35mmHg
Density: 1.568 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00075341
CAS Number: 86393-34-2
EINECS: 428-390-1
UN: 3265
ADR: 8,II