α,α,α,2,3,5,6-Heptafluorotoluene 98%
Supplier: Apollo Scientific
Warning
PC3424-25G PC3424-5G
APOSPC3424-25GEA 192 EUR
APOSPC3424-25G APOSPC3424-5G
α,α,α,2,3,5,6-Heptafluorotoluene 98%
α,α,α,2,3,5,6-Heptafluorotoluene
Formula: C₇HF₇
MW: 218.07 g/mol
Boiling Pt: 111…112 °C
Density: 1.601 g/cm³ (25 °C)
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00012385
CAS Number: 651-80-9
UN: 1993
ADR: 3,III